Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aluminum fluoride hydrate, Puratronic™, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1062306

Quantity 5g

 View more versions of this product

Catalog No. AA1062306


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

32287-65-3
MFCD00003426
Aluminum fluoride hydrate
83.98 (Anhydrous)
UN3260
Puratronic™

Specifications

32287-65-3
83.98 (Anhydrous)
UN3260
5g
99.99%, (metals basis)
Powder
MFCD00003426
−22 Mesh
Aluminum fluoride hydrate
Puratronic™

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit