Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Fluoride, Powder, 60%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 kg
Packaging:
Drum
Poly Bottle
Poly Pail
15098-87-0
15098-87-0
Grade Reagent
CAS Max % 1
Percent Purity 0.6
Packaging Poly Pail
Quantity 2.5 kg

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.