Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Foil, 0.5mm (0.02 in.) thick, Puratronic™, 99.998% (Metals basis), Alfa Aesar™

Form: Approx. 3.4g/50 x 50mm; Temper: as rolled

Manufacturer:  Alfa Aesar 11374HP

Quantity 100 x 500mm

 View more versions of this product

Catalog No. AA11374HP


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7429-90-5
99.998% (Metals basis)
Aluminum
Foil, 0.5mm (0.02 in.) thick
MFCD00134029

Specifications

100 x 500mm
Silver-colored foil
99.998% (Metals basis)
Odorless
Aluminum
7429-90-5
Foil, 0.5mm (0.02 in.) thick
MFCD00134029
Silver
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit