Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aluminum foil, 1.0mm (0.04 in.) thick, annealed, 99.99% (metals basis)

Click to view available options
Quantity:
50 x 50mm
100 x 100mm
100 x 500mm
7429-90-5
Al
26.982
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminium,metal,powder,aluminio,metana,adom,alumina fibre,aluminium flake,dust,noral aluminium
5359268
CHEBI:33629
aluminum
[Al]
7429-90-5
Al
26.982
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminium,metal,powder,aluminio,metana,adom,alumina fibre,aluminium flake,dust,noral aluminium
5359268
CHEBI:33629
aluminum
[Al]
Quantity 100 x 500mm
Weight ≈6.75g/50x50mm
Odor Odorless
Physical Form Foil
Percent Purity 99.99%
Chemical Name or Material Aluminum foil, 1.0mm (0.04 in.) thick, Annealed, Temper: soft
Assay (metals basis)

EINECSNumber : 231-072-3

TSCA : Yes

Recommended Storage : Ambient temperatures