Alfa Aesar™ Aluminum foil, 2.0mm (0.08 in.) thick, annealed, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 40763FI

Quantity 50x50mm

 View more versions of this product

Catalog No. AA40763FI

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7429-90-5
    MDL Number MFCD00134029
    Chemical Name or Material Aluminum foil, 2.0mm (0.08 in.) thick, Annealed, Temper: soft

    Specifications

    MDL Number MFCD00134029
    Physical Form Foil
    Odor Odorless
    TSCA TSCA
    CAS 7429-90-5
    Weight ≈13.5g/50x50mm
    Quantity 50x50mm
    Chemical Name or Material Aluminum foil, 2.0mm (0.08 in.) thick, Annealed, Temper: soft
    Assay 99.99%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit