Alfa Aesar™ Aluminum foil, alloy 6061, 1.0mm (0.04 in.) thick

Manufacturer: Alfa Aesar™ 45192HB

Quantity 300x300mm

 View more versions of this product

Catalog No. AA45192HB

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00134029
    Chemical Name or Material Aluminum foil, Alloy 6061, 1.0mm (0.04 in.) thick

    Specifications

    TSCA TSCA
    Physical Form Foil
    MDL Number MFCD00134029
    Odor Odorless
    Quantity 300x300mm
    Chemical Name or Material Aluminum foil, Alloy 6061, 1.0mm (0.04 in.) thick

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit