Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aluminum ICP Standard, 10,000 ppm Al in 5% HNO3, Ricca Chemical
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
Specifications
Physical Form | Liquid |
CAS Min % | 81.0, 11.23, 6.06 |
CAS Max % | 84.0, 11.69, 6.31 |
Packaging | Natural Poly Bottle |
Equivalence | 1 mL = 10 mg Al |
Color | Colorless |
Quantity | 50 mL |
Concentration or Composition Notes | 10,000ppm Al |
Chemical Name or Material | Aluminum ICP Standard |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction