Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum ICP Standard, 1mL = 1mg Al (1,000ppm Al), Ricca Chemical

Click to view available options
Quantity:
100mL
500mL
Packaging:
Poly natural
7784-27-2,7697-37-2,7784-27-2,7732-18-5
AlH18N3O18
375.129
SWCIQHXIXUMHKA-UHFFFAOYSA-N
24567
aluminum;trinitrate;nonahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
7784-27-2,7697-37-2,7784-27-2,7732-18-5
AlH18N3O18
375.129
SWCIQHXIXUMHKA-UHFFFAOYSA-N
24567
aluminum;trinitrate;nonahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
CAS Min % 93.0,4.17,1.29
CAS Max % 96.0,4.34,1.34
Physical Form Liquid
Packaging Poly natural
Concentration or Composition Notes 1,000ppm Al
Equivalence 1 mL = 1 mg Al
Color Colorless
Quantity 100mL
Chemical Name or Material Aluminum ICP Standard