Aluminum isopropoxide, 98+%

Manufacturer: Alfa Aesar 14007A1

Quantity 1kg

 View more versions of this product

Catalog No. AA14007A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 555-31-7
    Molecular Formula Al[OCH(CH3)2]3
    Formula Weight 204.25
    MDL Number MFCD00008870
    UN Number UN3181
    Merck Index 14,347
    Chemical Name or Material Aluminum isopropoxide

    Specifications

    Formula Weight 204.25
    Quantity 1kg
    TSCA TSCA
    MDL Number MFCD00008870
    UN Number UN3181
    Merck Index 14,347
    Solubility Information Soluble in alcohol, benzene, and toluene; Decomposes in water
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H228
    Flammable solid.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
    RTECS Number BD0975000
    Physical Form Granular
    Density 1.035
    Molecular Formula Al[OCH(CH3)2]3
    Storage Note 1 Moisture Sensitive
    Assay Percent Range 98+%
    CAS 555-31-7
    Chemical Name or Material Aluminum isopropoxide
    Assay ≥98%

    GHS H Statement
    H228
    Flammable solid.

    GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.

    Moisture Sensitive

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit