Learn More
Brady™ Aluminum Lockout Tagout Sign: THIS MACHINE MUST BE LOCKED OUT PRIOR TO SERVICING. FAILURE TO LOCK OUT MAY RESULT IN INJURY .
Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to clearly mark lockout and tagout areas.
Manufacturer:  Brady™ 144513
 View more versions of this product
Catalog No. 19-110-681
Add to cart
Description
Description
  • Aluminum material is abrasion-resistant and chemical-resistant
  • Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • For indoor and outdoor use
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
Specifications
Specifications
Black/yellow on white
Aluminum
Rectangle
Corner holes
230°F
-40°F
Non-adhesive
Not Laminated
1/Ea.
7 in. x 10 in. x 0.035 in.
10 in.
THIS MACHINE MUST BE LOCKED OUT PRIOR TO SERVICING. FAILURE TO LOCK OUT MAY RESULT IN INJURY .
0.035 in.
7 in.