Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aluminum Magnesium gauze , alloy 5056, 16 mesh woven from 0.24mm (0.0095 in.) dia. wire, Thermo Scientific™
$78.65
Specifications
Chemical Name or Material | Aluminum Magnesium gauze, Alloy 5056 |
---|---|
Name Note | 16 mesh woven from 0.24mm (0.0095 in.) dia. wire |
Avg. Mol. Wt. or Mol. Wt. Range | Al:Mg; 95:5 wt% |
MDL Number | MFCD00214039 |
Mesh Size | 16 Mesh |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA46714GT
|
Thermo Scientific Chemicals
46714GT |
150 x 150 mm |
Each of 1
for $78.65
Save 15% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Aluminum Magnesium gauze, Alloy 5056 | |
Al:Mg; 95:5 wt% | |
16 Mesh | |
Wire Cloth |
16 mesh woven from 0.24mm (0.0095 in.) dia. wire | |
MFCD00214039 | |
Odorless |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title