Alfa Aesar™ Aluminum magnesium isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol

Manufacturer: Alfa Aesar™ 36591AE

Quantity 100mL

 View more versions of this product

Catalog No. AA36591AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 69207-83-6
    Formula Weight 550.53
    MDL Number MFCD00070426
    UN Number UN1219
    Synonym Magnesium aluminum isopropoxide
    Chemical Name or Material Aluminum magnesium isopropoxide

    Specifications

    Formula Weight 550.53
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Concentration 10% w/v in isopropanol
    Quantity 100mL
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H336
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.
    Sensitivity Moisture Sensitive
    MDL Number MFCD00070426
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
    Boiling Point 130° to 142°C (2.0mmHg)
    Physical Form Liquid
    UN Number UN1219
    Color Colorless
    CAS 69207-83-6
    Synonym Magnesium aluminum isopropoxide
    Chemical Name or Material Aluminum magnesium isopropoxide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H225-H315-H319-H336
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit