Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum magnesium isopropoxide, 99.95+% (metals basis), Thermo Scientific Chemicals

Catalog Number aa3932004
Click to view available options
Quantity:
2 g
10 g
50 g
69207-83-6
MFCD00070426
69207-83-6
MFCD00070426

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 40°C
Boiling Point 114°C (0.2 mmHg)
Quantity 2 g
UN Number UN3181
Sensitivity Moisture sensitive
Formula Weight 550.53
Percent Purity ≥99.95%
Physical Form Crystalline Low Melting Solid
Assay (metals basis)
Chemical Name or Material Aluminum magnesium isopropoxide
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H225-H228-H315-H319-H335
Highly flammable liquid and vapor.
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit