Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Monostearate, NF, 14.7-16.7%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7047-84-9
7047-84-9

AL228, 7047-84-9

Spectrum™ Aluminum Monostearate, NF , also known as Dihydroxyaluminum stearate, is used to form pharmaceutical gels. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Arsenic (As) 4 ppm
Percent Purity 14.7 to 16.7%
Loss on Drying 0.03
Packaging Poly Pail
Quantity 500 g