Learn More
MilliporeSigma™ Supelco™ Aluminum Multi-Plate Racks
For use with Latch-Lid™ Developing Chambers
Supplier: MilliporeSigma™ Supelco™ Z2660431EA
Specifications
Aluminum | |
Multi-plate Rack | |
Latch-Lid™ Developing Chambers | |
10 x 10 cm Plates |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Latch-Lid is a trademark of General Glassblowing, Inc.