Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aluminum nickel isopropoxide, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 4237509

Quantity 10g

 View more versions of this product

Catalog No. AA4237509


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

70504-57-3
MFCD00143593
Aluminum nickel isopropoxide
584.92
UN3181

Specifications

70504-57-3
Dark
Powder
MFCD00143593
Aluminum nickel isopropoxide
10g
Moisture Sensitive
584.92
UN3181
99.9%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H228-H351-H315-H319-H317-H335
Highly flammable liquid and vapor.
Flammable solid.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit