Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aluminum-nickel, Raney-type Alloy, Powder Al-Ni 50/50, ACROS Organics™
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Specifications
Quantity | 2.5 kg |
Density | 3.4600g/mL |
Fieser | 01,723; 02,293; 05,570; 06,502; 07,312; 01,723; 02,293; 05,570; 06,502; 07,312; 08,433; 09,405; 10,339; 11,457; 12,422; 13,265; 14,270; 15,278; 17,296 |
Nickel (Ni) | 48.0 to 53.0% |
Packaging | Glass bottle |
Formula Weight | 85.68 |
Physical Form | Powder |
Color | Black-Gray to Gray |
Melting Point | 1350.0°C |
Chemical Name or Material | Aluminium-nickel |
Safety and Handling
Danger