Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum-nickel, Raney-type Alloy, Powder Al-Ni 50/50, ACROS Organics™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
12635-29-9
AlNi
85.675
NPXOKRUENSOPAO-UHFFFAOYSA-N
aluminium-nickel,nickel aluminum,nickel-aluminum alloy,aluminum-nickel alloy,nickel-aluminum,aluminum-nickel catalyst,al.ni,aluminum, compd. with nickel 1:1,aluminum-nickel,aluminium nickel
6336846
aluminum;nickel
[Al].[Ni]
12635-29-9
AlNi
85.675
NPXOKRUENSOPAO-UHFFFAOYSA-N
aluminium-nickel,nickel aluminum,nickel-aluminum alloy,aluminum-nickel alloy,nickel-aluminum,aluminum-nickel catalyst,al.ni,aluminum, compd. with nickel 1:1,aluminum-nickel,aluminium nickel
6336846
aluminum;nickel
[Al].[Ni]
Quantity 100 g
Density 3.4600g/mL
Fieser 01,723; 02,293; 05,570; 06,502; 07,312; 01,723; 02,293; 05,570; 06,502; 07,312; 08,433; 09,405; 10,339; 11,457; 12,422; 13,265; 14,270; 15,278; 17,296
Nickel (Ni) 48.0 to 53.0%
Packaging Glass bottle
Formula Weight 85.68
Physical Form Powder
Color Black-Gray to Gray
Melting Point 1350.0°C
Chemical Name or Material Aluminium-nickel