Learn More
Aluminum nitrate hydrate, Puratronic™, 99.999% (metals basis excluding Hg), Thermo Scientific Chemicals
Description
Aluminum nitrate hydrate is used to produce alumina for the preparation of insulating papers, and in the extraction of actinide elements. It is also used in the preparation of rare earth aluminates, in the heating elements of cathode ray tubes, as a precursor for sol-gel synthesis of alumina, and on transformer core laminates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 73.5°C |
Color | White |
Physical Form | Crystalline |
Quantity | 500 g |
Assay Percent Range | 99.999% (Metals basis excluding Hg) |
Solubility Information | Soluble in water,methanol,ethanol,and ethylene glycol. |
IUPAC Name | aluminium(3+) trinitrate |
Formula Weight | 212.99 (Anhydrous) |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
Hazard Category | H272-H315-H319 |
Precautionary Statement | P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c |
DOTInformation | DOT Class: 5.1, Packaging Group: III |
EINECSNumber | 236-751-8 |
RTECSNumber | BD1050000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.