Aluminum nitrate nonahydrate, 98%, Alfa Aesar

Manufacturer: Alfa Aesar 12360A7

Quantity 5kg

 View more versions of this product

Catalog No. AA12360A7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7784-27-2
    Physical Form Crystalline
    Assay Percent Range 98%
    Molecular Formula Al(NO3)3·9H2O
    Formula Weight 375.13
    MDL Number MFCD00003419
    Chemical Name or Material Aluminum Nitrate Nonahydrate, Nonahydrate

    Specifications

    Formula Weight 375.13
    Quantity 5kg
    Melting Point 73.5°C
    UN Number UN 1438, White
    Color White
    DOT Information DOT Class: 5.1, Packaging Group: III
    EINECS Number 236-751-8
    Physical Form Crystalline
    Odor Odorless
    Assay Percent Range 98%
    Molecular Formula Al(NO3)3·9H2O
    Appearance White crystalline
    MDL Number MFCD00003419
    CAS 7784-27-2
    Health Hazard 1 H272-H315-H319-H303
    Storage Note 1 Hygroscopic
    Chemical Name or Material Aluminum Nitrate Nonahydrate, Nonahydrate

    Hygroscopic

    H272-H315-H319-H303

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit