Aluminum Nitrate Nonahydrate (Cryst./Certified ACS), Fisher Chemical

Packaging Glass Bottle
Quantity 250g

 View more versions of this product

Catalog No. A586-250

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 7784-27-2
    Physical Form Solid
    Assay Percent Range 98.0 to 102.0 %
    Molecular Formula AlN3O9 · 9 H2O
    Linear Formula Al(NO3)3·9H2O
    Formula Weight 375.13
    MDL Number MFCD00149132
    Synonym Nitric Acid Aluminum Salt
    Chemical Name or Material Aluminum Nitrate Nonahydrate
    Assay 98.0 to 102.0%
    Grade Certified ACS Grade

    Specifications

    Formula Weight 375.13
    Melting Point 73°C
    Quantity 250g
    DOT Information DOT Class 5.1, : Oxidizer
    Viscosity [NULL]
    Physical Form Solid
    Color Clear
    pH 2.5 to 3.5
    Molecular Formula AlN3O9 · 9 H2O
    Health Hazard 3 Emergency Overview
    Oxidizer: Contact with combustible/organic material may cause fire. Hygroscopic. Use personal protective equipment. Keep away from clothing and other combustible materials. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

    NFPA

    Health:2
    Flammability:1
    Instability:2
    Heavy Metals (as Pb) 0.001% max.
    Iron (Fe) 0.002% max.
    Sodium (Na) 0.005% max.
    Magnesium (Mg) 0.001% max.
    Packaging Glass Bottle
    Assay Percent Range 98.0 to 102.0 %
    Linear Formula Al(NO3)3·9H2O
    Synonym Nitric Acid Aluminum Salt
    Chloride 0.001% max.
    Identification Pass Test
    Insoluble Matter 0.005% max.
    Potassium (K) 0.002% max.
    Sulfate 0.005% max.
    MDL Number MFCD00149132
    CAS 7784-27-2
    ChemAlert Storage Symbol Yellow
    Health Hazard 2 DANGER!
    Calcium (Ca) 0.005% max.
    Chemical Name or Material Aluminum Nitrate Nonahydrate
    Assay 98.0 to 102.0%
    Grade Certified ACS Grade
     

    DANGER!

    Emergency Overview
    Oxidizer: Contact with combustible/organic material may cause fire. Hygroscopic. Use personal protective equipment. Keep away from clothing and other combustible materials. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

    NFPA

    Health:2
    Flammability:1
    Instability:2

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit