Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Nitrate, Nonahydrate, Crystal, ACS, 98-102%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7784-27-2
7784-27-2
Grade ACS
CAS Max % 1
Percent Purity 98 to 102%
Calcium (Ca) 5e-05
Chloride 1e-05
Heavy Metals 1e-05
Insoluble Matter 5e-05
Iron (Fe) 2e-05
Magnesium (Mg) 1e-05
Packaging Amber Glass Bottle
Potassium (K) 2e-05
Sodium (Na) 5e-05
Sulfate 5e-05
Quantity 125 g
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.