Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aluminum nitride, 98+%

Manufacturer:  Alfa Aesar™ 4464514

Quantity 25g

 View more versions of this product

Catalog No. AA4464514

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

24304-00-5
40.99
14,353
AlN
MFCD00003429
Aluminum nitride

Specifications

25g
3.26g/mL
AlN
MFCD00003429
Ammonia-like
115m2/g
>2200°C
≥98%
24304-00-5
Powder
40.99
14,353
10 to 20nm APS
Pale gray
Aluminum nitride

Safety and Handling

GHS H Statement
H315-H319-H335

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a

Warning

RTECSNumber : BD1055000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit