Alfa Aesar™ Aluminum nitride, 98+%

Manufacturer: Alfa Aesar™ 44645-06

Quantity 5g

 View more versions of this product

Catalog No. AA4464506

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 24304-00-5
    Molecular Formula AlN
    Formula Weight 40.99
    MDL Number MFCD00003429
    Merck Index 14,353
    Chemical Name or Material Aluminum nitride

    Specifications

    Formula Weight 40.99
    CAS 24304-00-5
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for sev
    Molecular Formula AlN
    Merck Index 14,353
    Health Hazard 1 Warning
    Particle Size 10 to 20nm APS
    Surface Area 115m2/g
    RTECS Number BD1055000
    Quantity 5g
    Density 3.26g/mL
    Physical Form Powder
    MDL Number MFCD00003429
    Odor Ammonia-like
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Color Pale gray
    TSCA TSCA
    Melting Point >2200°C
    Chemical Name or Material Aluminum nitride
    Assay ≥98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for sev

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit