Aluminum Nitride Paste, Alfa Aesar™

Manufacturer: Alfa Aesar 43959A3

Quantity 2kg

 View more versions of this product

Catalog No. AA43959A3

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula AlN
    MDL Number MFCD00003429
    Chemical Name or Material Aluminium Nitride, Air set in 1-4 hours, heat cure in 2 hours at 200°C

    Specifications

    Quantity 2kg
    Color Gray
    Odor Odorless
    Molecular Formula AlN
    Appearance Gray paste
    MDL Number MFCD00003429
    Chemical Name or Material Aluminium Nitride, Air set in 1-4 hours, heat cure in 2 hours at 200°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit