Aluminum nitride Rod, Round, 10mm diameter, Alfa Aesar™

Manufacturer: Alfa Aesar 42998DO

Quantity 150mm

 View more versions of this product

Catalog No. AA42998DO

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 24304-00-5
    MDL Number MFCD00003429
    Chemical Name or Material Aluminum nitride

    Specifications

    Quantity 150mm
    Color White to pale yellow
    Odor Odorless
    MDL Number MFCD00003429
    Appearance White to pale yellow rod
    CAS 24304-00-5
    Chemical Name or Material Aluminum nitride

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit