Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum nitride Rod, Round, 22mm diameter, Alfa Aesar™

Manufacturer:  Alfa Aesar 43000DR

Quantity 200mm

 View more versions of this product

Catalog No. AA43000DR


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

24304-00-5
MFCD00003429
Aluminum nitride

Specifications

200mm
White to pale yellow rod
Odorless
Aluminum nitride
White to pale yellow
24304-00-5
MFCD00003429
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit