Aluminum nitride Rod, Round; Diameter (mm), 10, Alfa Aesar™

Manufacturer: Alfa Aesar 42998DM

Quantity 100mm

 View more versions of this product

Catalog No. AA42998DM

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 24304-00-5
    MDL Number MFCD00003429
    Chemical Name or Material Aluminum nitride Rod, Round; Diameter (mm), 10

    Specifications

    Quantity 100mm
    Odor Odorless
    TSCA TSCA
    Physical Form Rod
    MDL Number MFCD00003429
    CAS 24304-00-5
    RTECS Number BD1055000
    Density 3.26
    Color White to Pale Yellow
    Melting Point >2200°C
    Chemical Name or Material Aluminum nitride Rod, Round; Diameter (mm), 10

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit