Alfa Aesar™ Aluminum oxide, 99%

Manufacturer: Alfa Aesar™ 47171A3

Quantity 2kg

 View more versions of this product

Catalog No. AA47171A3

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1344-28-1
    Formula Weight 101.96
    MDL Number MFCD00003424
    Merck Index 14,356
    Chemical Name or Material Aluminum oxide

    Specifications

    Formula Weight 101.96
    Odor Odorless
    MDL Number MFCD00003424
    Merck Index 14,356
    Boiling Point 2980°C
    Color White
    Melting Point 2045°C
    Density 3.965g/mL
    Particle Size 32 to 63 Micron APS
    Physical Form Powder
    Refractive Index 1.768
    Surface Area 200m2/g
    CAS 1344-28-1
    TSCA TSCA
    Quantity 2kg
    RTECS Number BD1200000
    Chemical Name or Material Aluminum oxide
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit