Learn More
Aluminum oxide, 99%, Thermo Scientific Chemicals
Description
It is essential in protein extraction as a component in the preparation of tissue. Aluminum oxide is a source of aluminum in reactions, an abrasive agent, and as a refractory material. Furthermore, Aluminum oxide has a relatively high thermal conductivity of 30 Wm-1K-1. Also Aluminum oxide can act as a catalyst in a variety of reactions such as the Claus process and in the dehydration of alcohols to alkenes. It is an inorganic compound used as a grinding or blending agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 2980°C |
Melting Point | 2045°C |
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Merck Index | 14,356 |
Solubility Information | Insoluble in water. |
Formula Weight | 101.96 |
Percent Purity | 99% |
Odor | Odorless |
Show More |
Safety and Handling
EINECSNumber | 215-691-6 |
RTECSNumber | BD1200000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.