Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum oxide, activated, acidic, Brockmann Grade I, 58 angstroms, Alfa Aesar™

Click to view available options
Quantity:
250g
1kg
5kg
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminium oxide,aluminum oxide,alpha-alumina,fasertonerde,abramant,abramax,abrarex,abrasit,aloxite,alundum
9989226
dialuminum;oxygen(2-)
[O-2].[O-2].[O-2].[Al+3].[Al+3]
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminium oxide,aluminum oxide,alpha-alumina,fasertonerde,abramant,abramax,abrarex,abrasit,aloxite,alundum
9989226
dialuminum;oxygen(2-)
[O-2].[O-2].[O-2].[Al+3].[Al+3]

Acidic; S.A. 150m2/g

Solubility Information Insoluble in water.
Formula Weight 101.96
Physical Form -60 Mesh Powder, S.A. 150m2/g
Grade Brockmann
Color White
Melting Point 2045°C
Quantity 250g
Chemical Name or Material Aluminum oxide, Brockmann Grade I

EINECSNumber : 215-691-6

TSCA : Yes

Recommended Storage : Ambient temperatures