Alfa Aesar™ Aluminum oxide, activated, catalyst support, 99% (metals basis)

Manufacturer: Alfa Aesar™ 31268-A1

Quantity 1kg

 View more versions of this product

Catalog No. AA31268A1

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1344-28-1
    Formula Weight 101.96
    MDL Number MFCD00003424
    Chemical Name or Material Aluminum oxide, Activated, catalyst support

    Specifications

    Formula Weight 101.96
    Particle Size 3.2mm
    RTECS Number BD1200000
    TSCA TSCA
    Odor Odorless
    Physical Form Spheres
    Surface Area 90m2/g
    MDL Number MFCD00003424
    CAS 1344-28-1
    Melting Point 2045°C
    Quantity 1kg
    Chemical Name or Material Aluminum oxide, Activated, catalyst support
    Assay 99%, (metals basis)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit