Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aluminum oxide, activated, catalyst support, 99% (metals basis)

Manufacturer:  Alfa Aesar™ 3126830

Quantity 250g

 View more versions of this product

Catalog No. AA3126830

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1344-28-1
101.96
MFCD00003424
Aluminum oxide, Activated, catalyst support

Specifications

1344-28-1
101.96
Odorless
90m2/g
250g
99%, (metals basis)
Spheres
MFCD00003424
3.2mm
2045°C
Aluminum oxide, Activated, catalyst support

Safety and Handling

RTECSNumber : BD1200000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit