Learn More
Aluminum oxide, activated, neutral, gamma-phase, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Aluminum oxide is used as an adsorbent, desiccating agent and in the manufacture of dental cements and refractories. It is used in protein extraction as a component in the preparation of tissues and in bushings, spindles, lapping, polishing, refractory coating for investment casting shells, decorative non-skid tile and pressure blasting. It is also used in varnishes, for textile impregnation, in cosmetics and as an intermediate in pharmaceutical production. It is a source of aluminum in various reactions. It is used as a catalyst in a variety of reactions such as the Claus process and in the dehydration of alcohols to alkenes and as a grinding or blending agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99.9% (Metals basis) |
Merck Index | 14,356 |
Solubility Information | Insoluble in water. |
IUPAC Name | dialuminium(3+) trioxidandiide |
Formula Weight | 101.96 |
Percent Purity | 99.9% |
Odor | Odorless |
Show More |
Safety and Handling
EINECSNumber | 215-691-6 |
RTECSNumber | BD1200000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.