Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum oxide, Alfa Aesar™, NanoArc™

30% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion w/ dispersant

Manufacturer:  Alfa Aesar 4540636

Quantity 500g

 View more versions of this product

Catalog No. AA4540636

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1344-28-1
Al2O3
MFCD00003424
Dry Powder
101.96
Aluminum oxide, Sold in collaboration with Nanophase Technologies Corporation

Specifications

1344-28-1
Al2O3
Off-white powder
Off-white
Aluminum oxide, Sold in collaboration with Nanophase Technologies Corporation
Dry Powder
101.96
MFCD00003424
500g

Safety and Handling

H226-H319

DOTInformation : DOT Class: 3, Packaging Group: III

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit