Aluminum oxide, Alfa Aesar™, NanoArc™

30% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion w/ dispersant

Manufacturer: Alfa Aesar 4540622

Quantity 100g

 View more versions of this product

Catalog No. AA4540622

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1344-28-1
    Physical Form Dry Powder
    Molecular Formula Al2O3
    Formula Weight 101.96
    MDL Number MFCD00003424
    Chemical Name or Material Aluminum oxide, Sold in collaboration with Nanophase Technologies Corporation

    Specifications

    Physical Form Dry Powder
    Formula Weight 101.96
    Quantity 100g
    Color Off-white
    DOT Information DOT Class: 3, Packaging Group: III
    Density 1.2-1.8g/cm3
    Molecular Formula Al2O3
    Appearance Off-white powder
    MDL Number MFCD00003424
    CAS 1344-28-1
    Health Hazard 1 H226-H319
    Chemical Name or Material Aluminum oxide, Sold in collaboration with Nanophase Technologies Corporation

    H226-H319

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit