Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Oxide, Basic, Brockmann Activity Grade I, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

1344-28-1
Al2O3
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226

High quality chemicals for laboratory and specialized industrial use

    For Chromatography
SMILES [O-2].[O-2].[O-2].[Al+3].[Al+3]
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Physical Form Powder
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 4.00kg
Quantity 500g
Chemical Name or Material Aluminum Oxide, Basic, Brockmann Activity Grade I
Grade BAKER ANALYZED™ Reagent