Alfa Aesar™ Aluminum oxide, for Bio-Mass Cleanup

For Bio-Mass Cleanup

Manufacturer: Alfa Aesar™ 4620030

Quantity 250g

 View more versions of this product

Catalog No. AA4620030

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1344-28-1
    Molecular Formula Al2O3
    Linear Formula Al2O3
    Formula Weight 101.96
    MDL Number MFCD00003424
    Merck Index 14,356
    Chemical Name or Material Aluminum oxide

    Specifications

    Formula Weight 101.96
    Merck Index 14,356
    TSCA TSCA
    Odor Odorless
    Physical Form Powder
    Molecular Formula Al2O3
    Linear Formula Al2O3
    Boiling Point 2980°C
    Color White
    Particle Size 50 to 150 Micron APS
    Refractive Index 1.765
    MDL Number MFCD00003424
    CAS 1344-28-1
    Melting Point 2045°C
    Quantity 250g
    Chemical Name or Material Aluminum oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit