Learn More
Aluminum oxide, catalyst support, high surface area, 1/4in. Rings, Alfa Aesar™
S.A. 150 m2/g, medium pore diameter 130Å, total pore volume 0.77cc/g
Manufacturer:  Alfa Aesar 43858A1
Quantity 1kg
 View more versions of this product
Catalog No. AA43858A1
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
MFCD00003424
Aluminum oxide, catalyst support, high surface area, 1/4in. Rings
Specifications
MFCD00003424
1kg
Aluminum oxide, catalyst support, high surface area, 1/4in. Rings