Alfa Aesar™ Aluminum oxide, Drysphere™ desiccant, with 20% indicator

Manufacturer: Alfa Aesar™ 4612630

Quantity 250g

 View more versions of this product

Catalog No. AA4612630

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 1344-28-1
    Molecular Formula Al2O3
    Formula Weight 101.96
    MDL Number MFCD00003424
    Merck Index 14,356
    Chemical Name or Material Aluminum oxide, Drysphere™ desiccant, With 20% indicator

    Specifications

    Formula Weight 101.96
    Physical Form Spheres
    Molecular Formula Al2O3
    Boiling Point 2980°C
    Odor Odorless
    Particle Size 3.175mm
    Refractive Index 1.765
    CAS 1344-28-1
    Color White
    Melting Point 2045°C
    Quantity 250g
    TSCA TSCA
    MDL Number MFCD00003424
    Merck Index 14,356
    Chemical Name or Material Aluminum oxide, Drysphere™ desiccant, With 20% indicator

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit