Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum oxide, gamma-phase, 99.997% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4257622

Quantity 100g

 View more versions of this product

Catalog No. AA4257622

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1344-28-1
101.96
14,356
Al2O3
MFCD00003424
Aluminum oxide, gamma-phase

Specifications

1344-28-1
Al2O3
MFCD00003424
Odorless
2980°C
100g
99.997%, (metals basis)
110μm Powder
101.96
14,356
60m2/g
2045°C
Aluminum oxide, gamma-phase

Safety and Handling

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit