Aluminum oxide, gamma-phase, 99.997% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 42576-36

Quantity 500g

 View more versions of this product

Catalog No. AA4257636

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1344-28-1
    Molecular Formula Al2O3
    Formula Weight 101.96
    MDL Number MFCD00003424
    Merck Index 14,356
    Chemical Name or Material Aluminum oxide, gamma-phase

    Specifications

    Formula Weight 101.96
    Quantity 500g
    MDL Number MFCD00003424
    Melting Point 2045°C
    TSCA TSCA
    Odor Odorless
    Physical Form 110μm Powder
    Molecular Formula Al2O3
    Surface Area 60m2/g
    CAS 1344-28-1
    Merck Index 14,356
    Boiling Point 2980°C
    Chemical Name or Material Aluminum oxide, gamma-phase
    Assay 99.997%, (metals basis)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit