Alfa Aesar™ Aluminum oxide, gamma-phase, alpha-phase, 99.98% (metals basis)

Manufacturer: Alfa Aesar™ 1045914

Quantity 25g

 View more versions of this product

Catalog No. AA1045914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1344-28-1
    Formula Weight 101.96
    MDL Number MFCD00003424
    Merck Index 14,356
    Chemical Name or Material Aluminum oxide, gamma-phase, alpha-phase

    Specifications

    Formula Weight 101.96
    MDL Number MFCD00003424
    Merck Index 14,356
    TSCA TSCA
    Odor Odorless
    Physical Form Powder
    Surface Area 100m2/g
    Boiling Point 2980°C
    Density 3.965g/mL
    Particle Size 0.01 to 0.02 Micron
    CAS 1344-28-1
    Melting Point 2045°C
    Quantity 25g
    Chemical Name or Material Aluminum oxide, gamma-phase, alpha-phase
    Assay 99.98%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit