Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aluminum oxide, NanoDur™ AL-2420, 50% in mineral spirits, colloidal dispersion with dispersant, Thermo Scientific™
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Aluminum oxide dispersed in mineral spirits suitable for incorporation in non-polar organic based coatings and formulations
Specifications
Physical Form | Dry Powder |
Quantity | 100 g |
Formula Weight | 101.96 |
Concentration or Composition (by Analyte or Components) | 50% in mineral spirits |
Odor | Mild |
Chemical Name or Material | Aluminum oxide, NanoDur™ AL-2420, Colloidal dispersion with dispersant |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement H304 May be fatal if swallowed and enters airways. |
Precautionary Statement | GHS P Statement P301+P310-P331-P405-P501a IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
TSCA | TSCA |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction