Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aluminum oxide, NanoDur™ X1130PMA, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion

Manufacturer:  Alfa Aesar™ 4492636

Quantity 500g

 View more versions of this product

Catalog No. AA4492636

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

  • Aluminum oxide dispersion in Dowanol™ PMA suitable for incorporation in a wide variety of coatings formulations
Description & Specifications

Chemical Identifiers

1344-28-1
101.96
UN3271
Al2O3
MFCD00003424
Aluminum oxide, NanoDur™ X1130PMA, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion

Specifications

1344-28-1
Al2O3
MFCD00003424
45nm APS
Off-white to gray
42°C (108°F)
Dry Powder (Typical)
101.96
UN3271
146°C
500g
Aluminum oxide, NanoDur™ X1130PMA, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit