Alfa Aesar™ Aluminum oxide, NanoDur™ X1130PMA, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion

Manufacturer: Alfa Aesar™ 4492622

Quantity 100g

 View more versions of this product

Catalog No. AA4492622

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1344-28-1
    Molecular Formula Al2O3
    Formula Weight 101.96
    MDL Number MFCD00003424
    UN Number UN3271
    Chemical Name or Material Aluminum oxide, NanoDur™ X1130PMA, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion

    Specifications

    Physical Form Dry Powder (Typical)
    Formula Weight 101.96
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 1344-28-1
    TSCA TSCA
    Particle Size 45nm APS
    Molecular Formula Al2O3
    Flash Point 42°C (108°F)
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
    MDL Number MFCD00003424
    Color Off-white to gray
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    UN Number UN3271
    Boiling Point 146°C
    Quantity 100g
    Chemical Name or Material Aluminum oxide, NanoDur™ X1130PMA, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion

    Description

    • Aluminum oxide dispersion in Dowanol™ PMA suitable for incorporation in a wide variety of coatings formulations

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit