Alfa Aesar™ Aluminum oxide, NanoTek™ AL-6021, 50% in H2O, colloidal dispersion with dispersant

Manufacturer: Alfa Aesar™ 45011-22

Quantity 100g

 View more versions of this product

Catalog No. AA4501122

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1344-28-1
    Molecular Formula Al2O3
    Formula Weight 101.96
    MDL Number MFCD00003424
    Chemical Name or Material Aluminum oxide, NanoTek™ AL-6021, Colloidal dispersion with dispersant

    Specifications

    Physical Form Dry Powder (Typical)
    Formula Weight 101.96
    MDL Number MFCD00003424
    TSCA TSCA
    Odor Odorless
    Particle Size 45nm APS
    Molecular Formula Al2O3
    CAS 1344-28-1
    Quantity 100g
    Concentration 50% in H2O
    Chemical Name or Material Aluminum oxide, NanoTek™ AL-6021, Colloidal dispersion with dispersant

    Description

    • Aluminum oxide water borne dispersion suitable for use in ultrafine acid side polishing slurries, designed to be especially suitable for marble polishing formulations

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit