Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aluminum oxide, NanoTek™ AL-6081, 23% in H2O, colloidal dispersion with dispersant

Manufacturer:  Alfa Aesar™ 4490736

Quantity 500g

 View more versions of this product

Catalog No. AA4490736

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

  • Aluminum oxide water borne dispersion suitable for use in ultrafine acid side polishing slurries
Description & Specifications

Chemical Identifiers

1344-28-1
Al2O3
MFCD00003424
Aluminum oxide, NanoTek™ AL-6081, Colloidal dispersion with dispersant

Specifications

1344-28-1
Al2O3
23% in H2O
500g
Dry Powder (Typical)
MFCD00003424
45nm APS
Aluminum oxide, NanoTek™ AL-6081, Colloidal dispersion with dispersant

Safety and Handling

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit