Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aluminum Oxide, Neutral, Brockmann Activity Grade I, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

1344-28-1
Al2O3
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226

High quality chemicals for laboratory and specialized industrial use

    For Chromatography
CAS 1344-28-1
Molecular Formula Al2O3
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES [O-2].[O-2].[O-2].[Al+3].[Al+3]
IUPAC Name dialuminum;oxygen(2-)
Molecular Weight (g/mol) 101.96
PubChem CID 9989226
Formula Weight 101.96
Physical Form Powder
Packaging Poly Bottle
Specific Gravity 1L = 4.00kg
Quantity 500g
Chemical Name or Material Aluminum Oxide, Neutral, Brockmann Activity Grade I
Grade BAKER ANALYZED™ Reagent
Show More Show Less