Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aluminum Oxide, Neutral, Brockmann Activity Grade I, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Description
- For Chromatography
Specifications
Specifications
Physical Form | Powder |
Quantity | 500 g |
IUPAC Name | dialuminum;oxygen(2-) |
Formula Weight | 101.96 |
Grade | BAKER ANALYZED™ Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 4.00kg |
Chemical Name or Material | Aluminum Oxide, Neutral, Brockmann Activity Grade I |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction